missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP120B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94561-0.1ml
This item is not returnable.
View return policy
Description
TIP120B Polyclonal antibody specifically detects TIP120B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TIP120B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 850-1000 of human CAND2 (NP_001155971.1). VGQVAGPGHQRELKAVLLEALGSPSEDVRAAASYALGRVGAGSLPDFLPFLLEQIEAEPRRQYLLLHSLREALGAAQPDSLKPYAEDIWALLFQRCEGAEEGTRGVVAECIGKLVLVNPSFLLPRLRKQLAAGRPHTRSTVITAVKFLISD | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23066 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction