missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIRAP Antibody (20D1055.1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56730
This item is not returnable.
View return policy
Description
TIRAP Monoclonal specifically detects TIRAP in Human samples. It is validated for Western Blot.
Specifications
| TIRAP | |
| Monoclonal | |
| Unconjugated | |
| P58753 | |
| TIRAP | |
| This antibody was developed against a synthetic peptide corresponding to a portion of human TIRAP. | |
| 0.2 mL | |
| Primary | |
| In HUVEC, a 32 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM κ |
| Western Blot | |
| 20D1055.1 | |
| Western Blot 1:500-1:1000 | |
| adapter protein wyatt, Adaptor protein Wyatt, MAL, MyD88 adapter-like protein, MyD88-2, TIR domain-containing adapter protein, toll/interleukin-1 receptor domain-containing adapter protein, toll-interleukin 1 receptor (TIR) domain containing adaptor protein, Toll-interleukin 1 receptor (TIR) domain-containing adaptor protein, Toll-like receptor adaptor protein, wyatt | |
| Mouse | |
| Unpurified | |
| RUO | |
| 114609 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction