missing translation for 'onlineSavingsMsg'
Learn More

Tirofiban, MedChemExpress

Product Code. 16497308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16497308

Brand: Medchem Express HY17369B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tirofiban (L700462) is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist that inhibits fibrinogen binding to this receptor and has antithrombotic activity.

Tirofiban induces proliferation and migration on endothelial cell by inducing production of VEGF. Tirofiban can significantly reduces myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.

Specifications

Chemical Name or Material Tirofiban
CAS 144494-65-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H36N2O5S
Quantity 10 mg
Synonym L700462 MK383
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming) (insoluble or slightly soluble) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=S(N[C@H](C(O)=O)CC1=CC=C(OCCCCC2CCNCC2)C=C1)(CCCC)=O
Molecular Weight (g/mol) 440.6
Formula Weight 440.6
Percent Purity 98.37%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.