missing translation for 'onlineSavingsMsg'
Learn More

Tiron™

Product Code. 10052060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10052060 100 g Glass bottle 100g
10552581 25 g Glass bottle 25g
11321572 25 g Glass bottle 25g
10378793 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10052060 Supplier Thermo Scientific Acros Supplier No. 174141000
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149-45-1 | C6H10Na2O8S2 | 320.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
MDL Number MFCD00149531
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym 4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Melting Point >300.0°C
Infrared Spectrum Authentic
Loss on Drying 4% to 6% (typically)
Packaging Glass bottle
Linear Formula (HO)2C6H2(SO3Na)2·H2O
Beilstein 11, 297
Merck Index 15, 9621
Color White
Solubility Information Solubility in water: freely soluble.
Quantity 100 g
Physical Form Powder
Formula Weight 332.21
Chemical Name or Material Tiron™
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.