accessibility menu, dialog, popup

UserName

NaNaOHHOSOOOSOOO

CAS RN 149-45-1

IUPAC Name: disodium 4,5-dihydroxybenzene-1,3-disulfonate
Synonyms: tiron
Molecular Weight (g/mol): 314.19
Molecular Formula: C6H4Na2O8S2
InChi Key: ISWQCIVKKSOKNN-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=CC(=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Color 
  • (4)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results