missing translation for 'onlineSavingsMsg'
Learn More

Tiron™

Product Code. 10552581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10552581 25 g Glass bottle 25g
10052060 100 g Glass bottle 100g
10378793 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10552581 Supplier Thermo Scientific Acros Supplier No. 174140250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 149-45-1 | C6H10Na2O8S2 | 320.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
MDL Number MFCD00149531
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym 4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Melting Point >300.0°C
Linear Formula (HO)2C6H2(SO3Na)2·H2O
Beilstein 11, 297
Merck Index 15, 9621
Solubility Information Solubility in water: freely soluble.
Infrared Spectrum Authentic
Formula Weight 332.21
Loss on Drying 4% to 6% (typically)
Packaging Glass bottle
Color White
Quantity 25 g
Physical Form Powder
Chemical Name or Material Tiron™
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Dihydroxybenzene-3,5-disulfonic acid, disodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Dihydroxybenzene-3,5-disulfonic acid, disodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.