missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tissue alpha-L-Fucosidase/FUCA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
113.00 € - 363.00 €
Specifications
| Antigen | Tissue alpha-L-Fucosidase/FUCA1 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642594
|
Novus Biologicals
NBP3-21436-25ul |
25 μg |
113.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612565
|
Novus Biologicals
NBP3-21436-100ul |
100 μg |
363.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tissue alpha-L-Fucosidase/FUCA1 Polyclonal antibody specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for ELISASpecifications
| Tissue alpha-L-Fucosidase/FUCA1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2um filtered solution in PBS | |
| Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase | |
| Produced in rabbits immunized with purified, recombinant Human Tissue alpha-L-Fucosidase/FUCA1 (Gln32-Lys466; Uniprot: P04066) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P04066 | |
| 2517 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title