missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tissue Preservation Solution - HRP/AP assays

Product Code. 18664561 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18664561 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18664561 Supplier Novus Biologicals™ Supplier No. NBP312178
  • ONLINE EXCLUSIVE PRICE
    248.00 €
    234.15 € / 30mL
    Save 13.85 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Content And Storage Store at room temperature.
Quantity 30 mL
For Use With (Application) Immunohistochemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.