missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) iodide, anhydrous, 99.9% (metals basis)

Product Code. 11368588
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368588 50 g 50g
11378588 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368588 Supplier Thermo Scientific Alfa Aesar Supplier No. 040663.18
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7720-83-4

Titanium(IV) iodide, anhydrous is used in the CVD of titanium nitride films with optimized characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7720-83-4
MDL Number MFCD00043077

Specifications

Melting Point 150°C
Physical Form Lump
Quantity 50 g
UN Number UN3260
Solubility Information Soluble in dichloromethane,chloroform,carbon disulfide.
Formula Weight 555.52
Percent Purity 99.9%
Odor Acrid
Sensitivity Moisture sensitive
Density 4.30 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.