missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium Oil Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ti | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | Triple-leached LDPE bottle |
| CAS | 7440-32-6, 8042-47-5 |
| Certificate of Analysis | With Certificate of Analysis. |
| CAS Min % | 0.1, 99.9 |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| CAS Max % | 0.1, 99.9 |
| Molecular Formula | Ti |
| MDL Number | MFCD00011264,MFCD00151301 |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
| Show More |
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction