missing translation for 'onlineSavingsMsg'
Learn More

Titanium solution for ICP/MS 10mg/L HNO3/HF matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12967433 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12967433 100 mL 100mL
12957433 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12967433 Supplier Fisher Chemical Supplier No. S 661 100
  • ONLINE EXCLUSIVE PRICE
    64.65 €
    49.97 € / 100mL
    Save 14.68 €
    22% Off
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Nitric Acid/Trace Hydrogen Fluoride
Quantity 100 mL
Chemical Name or Material Titanium
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. (ICP & ICP-MS). Titanium (Ti) 10mg/l in HNO3 2% HF 0.2%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.