Learn More
DiversifiedBiotech Titer Tops with 100 Pk.
Product Code. 30282270
Change view
Click to view available options
Quantity:
100/Pk.
50/Pk.
Unit Size:
Pack of 100
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30282270 50/Pk. Pack of 50
30282264 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30282270 Supplier DiversifiedBiotech Supplier No. TTOPS50
  • ONLINE EXCLUSIVE PRICE
    55.20 €
    51.81 € / Pack of 50
    Save 3.39 €
    6% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pre-cut microplate film with polyethylene material
  • Pre-cut feature
  • Pierceable polyethylene microplate film for preventing cross-well contamination
  • End tabs make film positioning and removal hassle-free
  • Titer Tops with 100 Pk.
Specifications
Color Clear
Material Polypropylene
Sterility Sterile
Autoclavable Autoclavable
Quantity 50/Pk.
Product Type Titer Tops Seals
Adhesive Acrylic
Pierceable Yes