missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
379.00 € - 554.00 €
Specifications
| Antigen | Titin |
|---|---|
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491282
|
Novus Biologicals
NBP1-88071-25ul |
25 μL |
379.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000314
|
Novus Biologicals
NBP1-88071 |
0.1 mL |
554.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titin Polyclonal specifically detects Titin in Human, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Titin | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cytoskeleton Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7273 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GTVSTSCYLAVQVSEEFEKETTAVTEKFTTEEKRFVESRDVVMTDTSLTEEQAGPGEPAAPYFITKPVVQKLVEGGSVVFGCQVGGNPKPHVYWKKSGVPLTTGYRYKVSYNKQTGECKLVISMTFADDAGEYT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| cardiomyopathy, dilated 1G (autosomal dominant), CMD1G, CMH9, CMPD4, Connectin, DKFZp451N061, EC 2.7.11.1, EOMFC, FLJ32040, FLJ34413, FLJ39564, FLJ43066, HMERF, LGMD2JFLJ26020, MPRM, Rhabdomyosarcoma antigen MU-RMS-40.14, titin, TMDFLJ26409 | |
| TTN | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title