missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1, Mouse anti-Human, Clone: TLE1/2051, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79779-20UG
This item is not returnable.
View return policy
Description
TLE1 Monoclonal specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TLE1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q04724 | |
| TLE1 | |
| Recombinant human TLE1 fragment (aa 175-338) (exact sequence is proprietary) | |
| 20 μg | |
| Signal Transduction | |
| 7088 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TLE1/2051 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction