missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Antibody (CL3573) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61621H
This item is not returnable.
View return policy
Description
TLE3 Monoclonal antibody specifically detects TLE3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| TLE3 | |
| Monoclonal | |
| HRP | |
| ESG3Enhancer of split groucho-like protein 3, ESGenhancer of split groucho 3, GRG3, HsT18976, KIAA1547FLJ39460, transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 3, homolog of Drosophila E(sp1), transducin-like enhancer protein 3 | |
| This antibody was developed using a recombinant protein derived from Q04726, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 7090 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL3573 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction