Learn More
TLR2 Antibody (CL10552) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21176MFV500
Product Code. 30307764
  • 770.00 € / 0.10mL
Description
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR2
Monoclonal
mFluor Violet 500 SE
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
This antibody was developed using a synthetic peptide derived from O60603, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cytokine Research, Immunology, Innate Immunity, Signal Transduction
7097
Store at 4⪚C in the dark.
IgG1
Western Blot
CL10552
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified