missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR2 Recombinant Rabbit Monoclonal Antibody (067), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
131.00 € - 326.00 €
Specifications
| Antigen | TLR2 |
|---|---|
| Clone | 067 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019946
![]() |
Invitrogen™
MA552129 |
100 Tests |
326.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019960
|
Invitrogen™
MA552130 |
25 Tests |
131.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR2 Recombinant Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Flow CytometrySpecifications
| TLR2 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.09% sodium azide | |
| CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a | |
| Recombinant Human TLR2/CD282 Protein (Lys 19-Arg587). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Tlr2 |
| 067 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| O60603 | |
| 7097 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title