Learn More
TLR3 Antibody (40C1285.6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 75 publications
Brand:  Novus Biologicals NBP2-24875SS
 View more versions of this product
Product Code. 18715643
  • 244.00 € / 25µg
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Purified
IgG1 κ