missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody (1138D), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7960B
This item is not returnable.
View return policy
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| TLR9 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1138D | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9, Leu26-Asp818, Accession # AAK29625 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 54106 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction