missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM9SF2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00 € - 481.00 €
Specifications
| Antigen | TM9SF2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617342
|
Novus Biologicals
NBP2-95189-0.02ml |
0.02 mL |
204.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679561
|
Novus Biologicals
NBP2-95189-0.1ml |
0.1 mL |
481.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TM9SF2 Polyclonal antibody specifically detects TM9SF2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| TM9SF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ26287, MGC117391, P76, transmembrane 9 superfamily member 2, transmembrane protein 9 superfamily member 2,76 kDa membrane protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-300 of human TM9SF2 (NP_004791.1). GPRRSGAFYLPGLAPVNFCDEEKKSDECKAEIELFVNRLDSVESVLPYEYTAFDFCQASEGKRPSENLGQVLFGERIEPSPYKFTFNKKETCKLVCTKTYHTEKAEDKQKLEFLKKSMLLNYQHHWIVDNMPVTWCYDVEDGQRFCNPGFPIGCYITDKGHAKDACVISSDFHERDTFYIFNHVDIKIYYHVVETGSMGARLVAAKLEPKSFKHTHIDKPDCSGPPMDISNKASGEIKIAYTYSVSFEEDDKIRWASRWDYILESMPHTHIQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9375 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title