missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM173/STING Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19851-1-AP-150UL
This item is not returnable.
View return policy
Description
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.Specifications
| TMEM173/STING | |
| Polyclonal | |
| Unconjugated | |
| TMEM173 | |
| hMITA, hSTING, Mediator of IRF3 activation, MITA, MPYS, NET23, STING, TMEM173, transmembrane protein 173 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340061, 498840, 72512 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1M391, Q3TBT3, Q86WV6 | |
| TMEM173 | |
| TMEM173/STING Fusion Protein Ag13921 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction