Learn More
TMEM173/STING Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19851-1-AP-20UL
 View more versions of this product
Product Code. 16859334
  • 145.00 € / 20µL
Description
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.
Specifications
Specifications
TMEM173/STING
Polyclonal
Unconjugated
TMEM173
hMITA, hSTING, Mediator of IRF3 activation, MITA, MPYS, NET23, STING, TMEM173, transmembrane protein 173
Rabbit
Antigen Affinity Chromatography
RUO
340061, 498840, 72512
-20°C
Liquid
Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
F1M391, Q3TBT3, Q86WV6
TMEM173
TMEM173/STING Fusion Protein Ag13921
20 μL
Primary
Rat, Human, Mouse