missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM55B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23992-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 4,5-bisphosphate. Does not hydrolyze phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-bisphosphate, inositol 3,5-bisphosphate, inositol 3,4-bisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4-monophosphate and phosphatidylinositol 3-monophosphate.Specifications
| TMEM55B | |
| Polyclonal | |
| Unconjugated | |
| PIP4P1 | |
| C14orf9, PtdIns 4,5 P2 4 Ptase I, TMEM55B, transmembrane protein 55B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219024, 90809 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TWL2, Q86T03 | |
| PIP4P1 | |
| TMEM55B Fusion Protein Ag21172 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction