missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM86B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-31684
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TMEM86B Polyclonal specifically detects TMEM86B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| TMEM86B | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q8N661 | |
| TMEM86B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MDAGKAGQTLKTHCSAQRPDVC | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| lysoplasmalogenase, MGC30208, transmembrane protein 86B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 255043 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido