missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM92 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00 € - 322.00 €
Specifications
| Antigen | TMEM92 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653168
|
Novus Biologicals
NBP3-12676-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646497
|
Novus Biologicals
NBP3-12676-50ul |
50 μg |
195.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM92 Polyclonal antibody specifically detects TMEM92 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| TMEM92 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ33318, transmembrane protein 92 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM92 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 162461 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title