missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEPAI Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41321
This item is not returnable.
View return policy
Description
TMEPAI Polyclonal specifically detects TMEPAI in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TMEPAI | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| PMEPA1 | |
| Antibody was raised against a 16 amino acid peptide near the center of human PMEPA1. The immunogen is located within amino acids 160 - 210 of PMEPA1. Amino Acid Squence: QLELNRESVRAPPNRT | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least four isoforms are known to exist; this antibody will detect all of the isoforms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry 2.5 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/mL | |
| prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
| Rabbit | |
| 27 kDa | |
| 0.1 mg | |
| Signal Transduction | |
| 56937 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction