Learn More
Invitrogen™ TMF1 Recombinant Rabbit Monoclonal Antibody (24GB1965)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557767
 View more versions of this product
Product Code. 30274877
  • 198.00 € / 20µL
Description
TMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMF1
Recombinant Monoclonal
Unconjugated
TMF1
7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1
Rabbit
Affinity Chromatography
RUO
7110
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1965
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P82094
TMF1
A synthesized peptide derived from human TMF (aa 1000-1093).
20 μL