missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMF1 Recombinant Rabbit Monoclonal Antibody (24GB1965)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557766
This item is not returnable.
View return policy
Description
TMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
Specifications
| TMF1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TMF1 | |
| 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7110 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1965 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82094 | |
| TMF1 | |
| A synthesized peptide derived from human TMF (aa 1000-1093). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction