missing translation for 'onlineSavingsMsg'
Learn More

TMP269, MedChemExpress

Product Code. p-200100351
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487508

Brand: Medchem Express HY18360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TMP269 is a novel and selective class IIa histone deacetylase (HDAC) inhibitor with IC50s of 157 nM, 97 nM, 43 nM and 23 nM for HDAC4, HDAC5, HDAC7 and HDAC9, respectively.

TMP269 is a novel and selective class IIa histone deacetylase (HDAC) inhibitor with IC50s of 157 nM, 97 nM, 43 nM and 23 nM for HDAC4, HDAC5, HDAC7 and HDAC9, respectively.

Specifications

Chemical Name or Material TMP269
Physical Form Solid
CAS 1314890-29-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H21F3N4O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 41 mg/mL (79.69 mM)
SMILES O=C(NCC1(C2=NC(C3=CC=CC=C3)=CS2)CCOCC1)C4=CC=CC(C5=NOC(C(F)(F)F)=N5)=C4
Molecular Weight (g/mol) 514.52
Formula Weight 514.52
Percent Purity 98.23%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.