missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS11A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94784-0.02ml
This item is not returnable.
View return policy
Description
TMPRSS11A Polyclonal antibody specifically detects TMPRSS11A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TMPRSS11A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11A, Transmembrane Protease, Serine 11A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-421 of human TMPRSS11A (NP_872412.3). SASFQPNLTVHITGFGALYYGGESQNDLREARVKIISDDVCKQPQVYGNDIKPGMFCAGYMEGIYDACRGDSGGPLVTRDLKDTWYLIGIVSWGDNCGQKDKPGVYTQVTYYRNWIASKTGI | |
| 0.02 mL | |
| Apoptosis, Cell Biology | |
| 339967 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction