missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94598-0.02ml
This item is not returnable.
View return policy
Description
TMPRSS7 Polyclonal antibody specifically detects TMPRSS7 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| TMPRSS7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| transmembrane protease serine 7, transmembrane protease, serine 7, type II transmembrane serine protease 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 478-717 of human TMPRSS7 (NP_001036040.2). HRIIGGTDTLEGGWPWQVSLHFVGSAYCGASVISREWLLSAAHCFHGNRLSDPTPWTAHLGMYVQGNAKFVSPVRRIVVHEYYNSQTFDYDIALLQLSIAWPETLKQLIQPICIPPTGQRVRSGEKCWVTGWGRRHEADNKGSLVLQQAEVELIDQTLCVSTYGIITSRMLCAGIMSGKRDACKGDSGGPLSCRRKSDGKWILTGIVSWGHGSGRPNFPGVYTRVSNFVPWIHKYVPSLL | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 344805 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction