missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMUB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86731-25ul
This item is not returnable.
View return policy
Description
TMUB1 Polyclonal specifically detects TMUB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TMUB1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| C7orf21, chromosome 7 open reading frame 21, DULP, Hepatocyte odd protein shuttling protein, HOPS, MGC5442, SB144, transmembrane and ubiquitin-like domain containing 1, transmembrane and ubiquitin-like domain-containing protein 1, Ubiquitin-like protein DULP, Ubiquitin-like protein SB144 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 83590 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TMUB1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ATDSMRGEAPGAETPSLRHRGQAAQPEPSTGFTATPPAPDSPQEPLVLRLKFLNDSEQVARAWPHDTIGSLKRTQFPGREQQVRLIYQGQLLGDDTQTLGSLHLPPNCVLHC | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction