missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-α/CD69/CD4/CD3 Mouse anti-Human, APC,FITC,PE,PerCP-Cy™,¢5.5, Clone: SK7 (also known as Leu-4),6401.1111,L78,SK3 (also known as Leu3a), BD FastImmune™
Mouse Monoclonal Antibody
Brand: BD Biosciences 337190
This item is not returnable.
View return policy
Description
TNF-α/CD69/CD4/CD3 Monoclonal antibody specifically detects TNF-α/CD69/CD4/CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
| TNF-α/CD69/CD4/CD3 | |
| Monoclonal | |
| APC/FITC/PE/PerCP-Cyanine5.5 | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| SK7 (also known as Leu-4), 6401.1111, L78, SK3 (also known as Leu3a) | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction