Learn More
TNF Rat anti-Mouse, PE, Clone: MP6-XT22, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554419
Description
The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF.
Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Recombinant mouse TNF
Intracellular Staining
Specifications
| TNF | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant mouse TNF | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| MP6-XT22 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.