missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RI/TNFRSF1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
169.00 € - 726.00 €
Specifications
| Antigen | TNF RI/TNFRSF1A |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Flow Cytometry, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18070149
|
Novus Biologicals
NBP1-97453-0.025MG |
0.025 mg |
169.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705363
|
Novus Biologicals
NBP1-97453-0.05mg |
0.05 mg |
428.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719573
|
Novus Biologicals
NBP1-97453-0.2mg |
0.2 mg |
726.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF RI/TNFRSF1A Polyclonal specifically detects TNF RI/TNFRSF1A in Human, Mouse, Rat, Porcine, Bovine, Canine, Drosophila, Hamster, Monkey, Rabbit, Sheep, Xenopus, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TNF RI/TNFRSF1A | |
| Western Blot 1:1000, Flow Cytometry 1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cytokine Research, Death Receptor Signaling Pathway | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 7132 | |
| Synthetic peptide corresponding to a portion of mouse TNF Receptor I. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Xenopus species, Rabbit, Pig, Human, Sheep, Bovine, Canine, Hamster, Monkey, Rat, Yeast, Drosophila, Mouse | |
| CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
| TNFRSF1A | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title