missing translation for 'onlineSavingsMsg'
Learn More

TNP, MedChemExpress

Product Code. 16494348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494348 10 mg 10mg
16460677 50 mg 50mg
16447067 5 mg 5mg
16425266 100 mg 100mg
16413046 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494348 Supplier Medchem Express Supplier No. HY110079
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 μM and 10.2 μM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes.

TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 μM and 10.2 μM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes.

Specifications

Chemical Name or Material TNP
Physical Form Solid
CAS 519178-28-0
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H16F3N7O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 125 mg/mL (281.93 mM)
SMILES FC(C1=CC(CNC2=NC(NCC3=CC=C([N+]([O-])=O)C=C3)=C4N=CNC4=N2)=CC=C1)(F)F
Molecular Weight (g/mol) 443.38
Formula Weight 443.38
Percent Purity 98.79%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.