missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tobramycin, 97%

Product Code. 15504870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15504870 5 g 5g
15512610 1 g 1g
15505156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15504870 Supplier Thermo Scientific Chemicals Supplier No. 455430050
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tobramycin, CAS# 32986-56-4, is an aminoglycoside antibiotic isolated from Streptomyces tenebrarius with a wide range of bacteriostatic activity, mainly against gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Tobramycin is an aminoglycoside compound derived from kanamycin B and displays antibiotic and bacteriostatic activity against gram-negative bacteria
• After entering the cells, it binds irreversibly to a specific aminoglycoside receptor on the bacterial 30S ribosomal subunit and interferes with the initiation complex between messenger RNA and the 30S subunit, consequently inhibiting the protein synthesis process leading to bacterial cell death
• Tobramycin also can interfere with protein elongation since it can induce misreading of the mRNA template, and cause incorrect amino acids to be incorporated into the growing polypeptide chain

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Type Tobramycin
Infrared Spectrum Authentic
Water 8% max.
Quantity 5 g
Solubility Information Solubility in water: soluble in cold water.
Formula Weight 467.51
Percent Purity 97%
Chemical Name or Material Tobramycin
compliance-icons
Product Identifier
  • Tobramycin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.