Learn More
Invitrogen™ Tocilizumab Recombinant Human Monoclonal Antibody (rhPM-1 (Tocilizumab))

Description
Specificity: Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response.
Specifications
Specifications
| Antigen | Tocilizumab |
| Applications | ELISA, Inhibition Assays |
| Classification | Recombinant Monoclonal |
| Clone | rhPM-1 (Tocilizumab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene Alias | Actemra; Atlizumab; MRA; R-1569; RHPM-1; RO-4877533 |
| Host Species | Human |
| Immunogen | The original antibody was generated by CDR grafting of mouse PM-1 antibody onto human REI and NEW framework regions to create rhPM-1. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.