Learn More
Invitrogen™ TOE1 Recombinant Rabbit Monoclonal Antibody (24GB1860)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557790
 View more versions of this product
Product Code. 30276469
  • 564.00 € / 100µL
Description
TOE1 Recombinant Monoclonal Antibody for Western Blot, Flow
Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TOE1
Recombinant Monoclonal
2.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96GM8, Q9D2E2
TOE1
A synthesized peptide derived from human TOE1 (aa 15-77).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1860
Unconjugated
TOE1
4930584N22Rik; 4933424D16Rik; AI413517; hCaf1z; Target of EGR1 protein 1; target of EGR1, member 1 (nuclear); TOE1
Rabbit
Affinity chromatography
RUO
114034, 298443, 68276
-20°C