missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tofacitinib citrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Description
Tofacitinib citrate is an orally available JAK1/2/3 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Tofacitinib citrate has antibacterial, antifungal and antiviral activities.
Specifications
Specifications
| Chemical Name or Material | Tofacitinib citrate |
| Physical Form | Solid |
| CAS | 540737-29-9 |
| Color | White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H28N6O8 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | Tasocitinib citrateCP-690550 citrate |
| Solubility Information | DMSO : 28.57 mg/mL (56.63 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (6.60 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction