missing translation for 'onlineSavingsMsg'
Learn More

Tolazamide, MedChemExpress

Product Code. 16464507
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464507 50 mg 50mg
16458638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16464507 Supplier Medchem Express Supplier No. HYB0920
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tolazamide is an oral blood glucose lowering drug used for people with Type 2 diabetes.

Tolazamide is an oral blood glucose lowering drug used for people with Type 2 diabetes.

Specifications

Chemical Name or Material Tolazamide
CAS 1156-19-0
Color White
Purity Grade Notes Research
Molecular Formula C14H21N3O3S
Quantity 50 mg
Synonym U-17835
Solubility Information DMSO : ≥ 39 mg/mL (125.24 mM)
SMILES O=S(C1=CC=C(C)C=C1)(NC(NN2CCCCCC2)=O)=O
Molecular Weight (g/mol) 311.4
Formula Weight 311.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.