missing translation for 'onlineSavingsMsg'
Learn More

Tolfenamic Acid, MedChemExpress

Product Code. 16488448
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488448 10 g 10g
16472139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488448 Supplier Medchem Express Supplier No. HYB0335
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tolfenamic Acid (GEA 6414) is a non-steroidal anti-inflammatory and anti-cancer agent, selectively inhibits COX-2, with an IC50 of 13.49 μM (3.53 μg/mL) in LPS-treated (COX-2) canine DH82 monocyte/macrophage cells, but shows no effect on COX-1.

Tolfenamic Acid (GEA 6414) is a non-steroidal anti-inflammatory and anti-cancer agent, selectively inhibits COX-2, with an IC50 of 13.49 μM (3.53 μg/mL) in LPS-treated (COX-2) canine DH82 monocyte/macrophage cells, but shows no effect on COX-1.

Specifications

Chemical Name or Material Tolfenamic Acid
Physical Form Powder
CAS 13710-19-5
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C14H12ClNO2
For Use With (Application) COVID-19-immunoregulation
Synonym GEA 6414
Solubility Information DMSO : ≥ 100 mg/mL (382.12 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C1=CC=CC=C1NC2=C(C)C(Cl)=CC=C2
Molecular Weight (g/mol) 261.7
Formula Weight 261.7
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.