missing translation for 'onlineSavingsMsg'
Learn More

Tolylene 2,4-Diisocyanate, TRC

Product Code. 30040959
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040959 50 g 50g
30040484 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30040959 Supplier Toronto Research Chemicals Supplier No. TRCT53671050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-84-9
Molecular Formula C9 H6 N2 O2
Molecular Weight (g/mol) 174.16
Synonym Isocyanic acid, 4-methyl-m-phenylene ester (8CI), 2,4-Diisocyanato-1-methylbenzene, 1,3-Diisocyanato-4-methylbenzene, 2,4-Diisocyanatotoluene, 2,4-TDI, 2,4-Toluene diisocyanate, 2,4-Toluylene diisocyanate, 2,4-Tolylene diisocyanate, 4-Methyl-1,3-phenylene diisocyanate, 4-Methyl-m-phenylene diisocyanate, 4-Methyl-m-phenylene isocyanate, NSC 4791, NSC 56759, TDI 100, Toluene 2,4-diisocyanate
IUPAC Name 2,4-diisocyanato-1-methylbenzene
SMILES Cc1ccc(cc1N=C=O)N=C=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3
Formula Weight 174.0429
Chemical Name or Material 2,4-Toluenediisocyanate (2,4-TDI)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.