missing translation for 'onlineSavingsMsg'
Learn More

TOMM40L Monoclonal Antibody (OTI6F3), TrueMAB™, OriGene

Product Code. 17504114
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17504114 30 μL 30µL
17584104 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17504114 Supplier Origene Technologies Supplier No. TA810705S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TOMM40L Monoclonal antibody specifically detects TOMM40L in Human samples. It is validated for Western Blot

Specifications

Antigen TOMM40L
Applications Western Blot
Classification Monoclonal
Clone OTI6F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FAM131C
Gene Accession No. Q96AQ9
Gene Alias hypothetical protein FLJ12770; mitochondrial import receptor subunit TOM40B; Protein TOMM40-like; RGD1562006; RP11-297K8.10; Tom40B; TOMM40B; Tomm40l; tomm40l {ECO:0000250; translocase of outer mitochondrial membrane 40 homolog (yeast)-like; translocase of outer mitochondrial membrane 40 homolog B; translocase of outer mitochondrial membrane 40 homolog, like; translocase of outer mitochondrial membrane 40 homolog-like; translocase of outer mitochondrial membrane 40 homolog-like (yeast); translocase of outer mitochondrial membrane 40 like; translocase of outer mitochondrial membrane 40B; UniProtKB:Q969M1}; zgc:85715
Host Species Mouse
Immunogen Full length human recombinant protein of human FAM131C produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348487
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.