missing translation for 'onlineSavingsMsg'
Learn More

Topotecan Hydrochloride, MedChemExpress

Product Code. 16455848
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455848 10 mg 10mg
16486316 100 mg 100mg
16451997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16455848 Supplier Medchem Express Supplier No. HY13768A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Topotecan Hydrochloride (SKF 104864A Hydrochloride) is a Topoisomerase I inhibitor with potent antineoplastic activities.

Topotecan Hydrochloride (SKF 104864A Hydrochloride) is a Topoisomerase I inhibitor with potent antineoplastic activities.

Specifications

Chemical Name or Material Topotecan Hydrochloride
CAS 119413-54-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H24ClN3O5
Quantity 10 mg
Synonym SKF 104864A HydrochlorideNSC 609669 Hydrochloride
Solubility Information DMSO : 100 mg/mL (218.38 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (72.79 mM; Need ultrasonic)
SMILES [H]Cl.O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=CC5=C(CN(C)C)C(O)=CC=C5N=C4C3=C2)=O
Molecular Weight (g/mol) 457.91
Formula Weight 457.91
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.