Learn More
Total Base Number Standard, ASTM D2896 (nominal value: 70.24 mg KOH/g), 3 x 50g, Paragon Scientific™
Description
- Base Number (TBN) tested and certified in accordance with ASTM D2896 / IP 276
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
- Typically used in, but not limited to, the analysis of used oils and lubricants
- Supplied as single volume packs or as triple packs
- Supplied in tamper-evident glass packaging, providing assurance of sample integrity
- Manufactured in the United Kingdom
- 24 month shelf life
Specifications
Specifications
| Quantity | 3 x 50 g |
Nominal values quoted. Certified Values will vary from batch to batch.