missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Total Inorganic Carbon (TIC) Standard 500 ppm, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Total Inorganic Carbon (TIC) Reference Standard, Total Inorganic Carbon (TIC) Reference Material.
- Independent, traceable and certified.
- Presented in a special twin-neck dosing device that prevents contamination, evaporation, or interference.
- Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
- Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
- The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
- The concentration of this standard is verified using a high performance calibrated total inorganic carbon analyzer (TIC Instrument).
- The calibration of the TIC instrument is completed using a high purity ISO Guide 34 accredited TIC standard similar in TIC concentration value to this product.
Specifications
Specifications
| Description | Accuracy ± 2&, Matrix: Water (H2O) |
| Packaging | HDPE Twin Neck Bottle |
| Concentration | 500 ppm |
| Technique | TIC Analysis |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction