missing translation for 'onlineSavingsMsg'
Learn More

Total Kjeldahl Nitrogen Standard: TKN @ 1000 mg/L in 1% HCl

Product Code. 30315813
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315813 100 mL 100mL
30311360 500 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30315813 Supplier VHG Labs Supplier No. VHGTOTKJN1K100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Total Kjeldahl Nitrogen
Solvent or Matrix Dilute HCl
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.