missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Total Organic Carbon Standard: TOC @ 1000 mg/L in H2O, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Total Organic Carbon Standard: TOC @ 1000 mg/L in H2O
Specifications
Specifications
| Chemical Name or Material | Carbon |
| CAS | 7440-44-0 |
| Methods | Wet Chemistry |
| Analyte or Component Names | Carbon |
| Shipping Condition | Room Temperature |
| Molecular Formula | C |
| Synonym | Carbon, Charcoal activated |
| InChI Formula | InChI=1S/C |
| SMILES | [C] |
| IUPAC Name | carbon |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction