missing translation for 'onlineSavingsMsg'
Learn More

Total Organic Carbon (TOC) Standard 5 ppm, Reagecon
BIOPHARMA

Product Code. 11536633 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11536633 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11536633 Supplier Reagecon Supplier No. TOC5
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    99.33 € / 500mL
    Save 22.67 €
    18% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from high-purity raw materials and suitable for calibrating a wide range of TOC analyzers to a high accuracy

  • Total Organic Carbon (TOC) Reference Standard, Total Organic Carbon (TOC) Reference Material.
  • Independent, traceable and certified.
  • Presented in a specially cleaned and leached amber glass bottle.
  • Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
  • Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
  • The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
  • The concentration of this standard is verified using a high performance calibrated total organic carbon analyzer (TOC Instrument).
  • The calibration of the TOC instrument is completed using a high purity ISO Guide 34 accredited TOC standard similar in TOC concentration value to this product.

Specifications

Description Accuracy ± 2&, Matrix: Water (H2O)
Packaging Amber Glass Bottle
Concentration 5 ppm
Technique TOC Analysis
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.